Learn More
Selleck Chemical LLC Combretastatin A4-10mg
SDP
Catalog No. 502389602
Encompass_Preferred
Combretastatin A4 is a microtubule-targeting agent that binds β-tubulin with Kd of 0.4 μM.Phase 3.
Catalog No. 50-238-9602 Supplier Selleck Chemical LLC Supplier No. S778310MG
Combretastatin A4 is a microtubule-targeting agent that binds β-tubulin with Kd of 0.4 μM.Phase 3.