Learn More
Enzo Life Sciences Comet SCGE assay kit 50tests
SDP
Catalog No. 50658192
Encompass_Preferred
Fast and simple electrophoresis method to measure DNA fragmentation. Assay Time: 3 hours. Applications: Fluorescence microscopy. Species Reactivity: Species independent. Contents: Lysis solution, Comet LMAgarose, Comet slide, EDTA, CYGREEN Nucleic acid dye
Catalog No. 50-658-192 Supplier Enzo Life Sciences Supplier No. ADI900166
Fast and simple electrophoresis method to measure DNA fragmentation. Assay Time: 3 hours. Applications: Fluorescence microscopy. Species Reactivity: Species independent. Contents: Lysis solution, Comet LMAgarose, Comet slide, EDTA, CYGREEN Nucleic acid dye