Learn More
MSC Comet Cleaner with Bleach, Disinfecting, 1 gal Jug, 3/Case
Catalog No. 502867195
Encompass
Comet Cleaner with Bleach is a powerful two-in-one liquid formula combining heavy-duty detergents with bleach for effective cleaning and stain removal. It is designed for various surfaces including restrooms and kitchens, acting as a hospital-use disinfectant when used per label instructions.
  • Combines cleaning power of heavy-duty detergents with bleach
  • Effective as a hospital-use disinfectant against Hepatitis A and C, Norovirus, and 39 other microorganisms
  • Kills 99.9% of bacteria and viruses
  • No abrasives
  • Suitable for ceramic, fiberglass, plastic, porcelain, stainless steel, and tile surfaces
  • Removes greasy soap scum, tracked-in dirt, scuffmarks, and stains
  • Included on the EPA List N for disinfectants to use against SARS-CoV-2
Catalog No. 50-286-7195 Supplier MSC Supplier No. 06901128
Comet Cleaner with Bleach is a powerful two-in-one liquid formula combining heavy-duty detergents with bleach for effective cleaning and stain removal. It is designed for various surfaces including restrooms and kitchens, acting as a hospital-use disinfectant when used per label instructions.
  • Combines cleaning power of heavy-duty detergents with bleach
  • Effective as a hospital-use disinfectant against Hepatitis A and C, Norovirus, and 39 other microorganisms
  • Kills 99.9% of bacteria and viruses
  • No abrasives
  • Suitable for ceramic, fiberglass, plastic, porcelain, stainless steel, and tile surfaces
  • Removes greasy soap scum, tracked-in dirt, scuffmarks, and stains
  • Included on the EPA List N for disinfectants to use against SARS-CoV-2