Learn More
R&D Systems™ CometAssay Lysis Solution

Description
- Store bottle at room temperature
- CometAssay™ Lysis Solution contains 1% sodium lauryl sarcosinate which is an irritant (precipitates with long term storage at 2-8 °C)
- In case of eye or skin contact, wash thoroughly under running water
- In case of ingestion, rinse mouth with water and seek medical advice
- CometAssay™ Lysis Solution contains high salt and detergent and is suitable for use with either the alkaline or neutral comet assay protocols
- Designed for use with CometAssay™, FLARE™, and CometChip™ Kits
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Isolation Technology | Alkaline Lysis |
| Formulation | Contains 1% sodium lauryl sarcosinate which is an irritant and precipitates with long term storage at 2-8°C. Contains high salt and detergent and is suitable for use with either the alkaline or neutral comet assay protocols. |
| For Use With (Equipment) | CometAssay™, FLARE™, & CometChip™ Kits |
| Product Type | Lysis Solution |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.