Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CometSlide™

Catalog No. 425000403
Change view
Click to view available options
Quantity:
2 Slides
25 Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-500-0403 2 Slides
42-500-5003 25 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-500-0403 Supplier R&D Systems™ Supplier No. 425000403
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sample-ready pre-treated glass slides

  • A component of the CometAssay™ Kit , which is used for the rapid analysis of DNA fragmentation associated with DNA damage
  • The CometAssay™, or single cell gel electrophoresis assay, permits the quantification of DNA damage in individual cells based on the migration of DNA structures in an electric field
  • Evaluation of the DNA Comet tail shape and migration pattern allows for assessment of DNA damage, expressed as tail length, percent DNA in the tail and tail moment
  • CometSlide™ provides a convenient, simple, and reliable format for sample preparation, processing, analysis, and storage for the CometAssay™

Specifications

Content And Storage Store the unopened product at room temperature. Do not use past expiration date.
Description Sample-ready pre-treated glass slides
For Use With (Equipment) CometAssay™ Kit
For Use With (Application) Sample preparation, processing, analysis, and storage for the CometAssay™
Quantity 2 Slides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.