Learn More
R&D Systems™ CometSlide™

Description
- A component of the CometAssay™ Kit , which is used for the rapid analysis of DNA fragmentation associated with DNA damage
- The CometAssay™, or single cell gel electrophoresis assay, permits the quantification of DNA damage in individual cells based on the migration of DNA structures in an electric field
- Evaluation of the DNA Comet tail shape and migration pattern allows for assessment of DNA damage, expressed as tail length, percent DNA in the tail and tail moment
- CometSlide™ provides a convenient, simple, and reliable format for sample preparation, processing, analysis, and storage for the CometAssay™
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Description | Sample-ready pre-treated glass slides |
| For Use With (Equipment) | CometAssay™ Kit |
| For Use With (Application) | Sample preparation, processing, analysis, and storage for the CometAssay™ |
| Product Line | R&D Systems |
| Product Type | Microscope Slide |
| Quantity | 25 Slides |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.