Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ 400 Series Replacement Harness
SDP

Catalog No. 19222564 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
400 series replacement harness; 12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-222-564 400 series replacement harness; 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-564 Supplier Dentec Safety™ Supplier No. 16132B12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Comfort-Air™ 400 series replacement harness includes eight adjustment points for a secure and comfortable fit.

  • Simple and easy to adjust and remove
  • Soft elastic strap feels great against the skin
  • Crown straps can be adjusted to accommodate pony tails and hair buns
  • Fits under face shields and welding helmets comfortably

Specifications

Color Black
Disposable Reusable
Material Nylon
Product Type Head Harness Assembly
Size One Size
For Use With (Equipment) Comfort-Air™ 400 series respirators
Latex Free Yes
Model 16132B12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.