Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 300 Half Mask Respirators
SDP

Catalog No. 19222413 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-413 Small/Medium
19-222-414 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-413 Supplier Dentec Safety™ Supplier No. 300SM00
Add to Cart
Add to Cart

Thermoplastic half-mask respirators provide comfort and durability with a flexible sealing edge and multi-position cartridge attachment.

  • Extended field of vision
  • Large inhalation valves
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Textured sealing area reduces risk of the mask slipping on the face to ensure a comfortable seal on a wide variety of face sizes
  • Enhanced harness assembly features:
    • Metal tabs at end to prevent nylon strap fraying and allowing for adjustment when wearing gloves
    • Wide nylon strap is more comfortable against the skin and maintains its elasticity longer
    • 4-point swiveling adjustments allows the mask to be adjusted
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Gray
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small/Medium
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Dentec™ Filters and Cartridges
Model 300-SM-00
Length (Metric) 8.89 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.