Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort Air™ Series 400 Low Maintenance Respirators
SDP

Catalog No. 19222416 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-416 Medium/Large
19-222-415 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-222-416 Supplier Dentec Safety™ Supplier No. 400ML00
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Elastomeric half-mask respirators features a flexible sealing edge and multi-position cartridge attachment.

  • Extended field of vision
  • Large inhalation valves
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Exhalation valve diaphragm has channels that act like a parachute to to exhaled air and lift the diaphragm off the valve seat with very little effort, creating more space for air to exit the mask reducing and thereby heat build up inside
  • Rolled sealing flange provides comfort and improves fitting capabilities
  • Textured sealing area reduces risk of the mask slipping on the face, ensuring a comfortable seal on a wide variety of facial sizes
  • Ultra light harness assembly provides additional comfort
  • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Dentec™ Filters and Cartridges
Model 400-ML-00
Length (Metric) 8.89 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.