Learn More
Dentec Safety™ Comfort-Air™ 400Nx Half-Mask Elastomeric Respirator
SDP
Catalog No. 19222551 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-551 Medium/Large
19-222-550 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-551 Supplier Dentec Safety™ Supplier No. 400NXML00
Add to Cart
Add to Cart
Comfort-Air™ 400Nx half-mask elastomeric respirator assemblies do not include cartridges. Can be used in conjunction with a N95 or P100 filter to provide protection against non-oil based particulate aerosols and COVID viruses.
  • Facepiece low profile design fits under industrial style and medical style face shields
    • Also provides extended field of vision
  • No exhalation valve ensures the user's exhaled breath does not exit the mask
  • Rolled sealing flange provides superior comfort and improves fitting capabilities
  • Does not require user to adjust straps excessively tight
  • Textured sealing area reduces risk of the mask slipping on the face
    • Comfortable seal on a wide variety of facial sizes
  • Designed to allow for the proper fitting of most safety & prescription eyewear and goggles
  • Ultra light harness assembly provides additional comfort
  • Cradle suspension fits comfortably under suspension systems for hard hats, grinding & welding helmets
  • 8-position adjustment allows you to adjust cartridges to three different positions
Specifications
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve No
Facepiece Material Thermoplastic Elastomer
Latex Free Yes
Model 400NX-ML-00
Suspension Type 8-point