Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Holder For N95 Filter Pads
SDP
Catalog No. 19222566 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Quantity:
350/Cs.
6 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-222-566 6 Pk.
19-222-567 350/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-566 Supplier Dentec Safety™ Supplier No. 14880W
Only null left
Add to Cart
Add to Cart
Dentec Comfort-Air™NxMD NxMD filter holder for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
  • Ultra thin design will fit under any grinding shield or welding helmet
  • Will not obstruct field of vision
  • Holder has spacers that create a space between the filter material and the holder
    • This allows the user to access the entire filter material increasing filter life while reducing breathing resistance and increasing productivity
Specifications
Color White
Disposable Reusable
Material Plastic
Product Type Filter Holder
With Exhalation Valve No
Latex Free Yes
Quantity 6 Pk.