Learn More
Dentec Safety™ Comfort-Air™ 400NxMD Half-Mask Elastomeric Respirator with Filter Cover & Holder
SDP
Catalog No. 19222559 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-559 Medium/Large
19-222-558 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-222-559 Supplier Dentec Safety™ Supplier No. 400NXMDMLNF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
White NxMD half-mask respirators feature a filter cover and holder and are NIOSH approved. OSHA silica dust compliant when used with a N95 or P100 filter. Provides protection against non-oil based particulate aerosols and COVID viruses.
  • Facepiece low-profile design fits under industrial style and medical style face shields
    • Extended field of vision
  • No exhalation valve ensures the user's exhaled breath does not exit the mask
  • Rolled sealing flange provides comfort and improves fitting capabilities
  • Does not require user to adjust straps excessively tight
  • Textured sealing area reduces risk of the mask slipping on the face
    • White color allows for written personalization and also makes dirt or other markings more visible to simplify the cleaning and disinfecting process
  • Designed to allow for the proper fitting of most safety & prescription eyewear and goggles
  • Ultra light harness assembly provides additional comfort
  • Cradle suspension fits comfortably under suspension systems for hard hats, grinding & welding helmets
  • 8-position adjustment
Specifications
Color White
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve No
Facepiece Material Thermoplastic Elastomer
Latex Free Yes
Model 400-NXMD-ML-NF
Suspension Type 8-point