Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 400 Elastomeric Facepiece Respirator with OV/N95 Filter Assembly
SDP

Catalog No. 19222470 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-222-470 Small/Medium
19-222-469 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-470 Supplier Dentec Safety™ Supplier No. 4SM20DN5ZAM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elastomeric half-mask respirators with N95 non-oil particulate filters feature a low-profile design that fit under most grinding and welding helmets.

  • Extended field of vision
  • Large inhalation valves
  • Complete with OV/N95 filter assembly for laquer and enamel paints, pesticides, welding, dusts and non-oil mist environments
  • Exhalation valve diaphragm has channels that act like a parachute to traps exhaled air and lift the diaphragm off the valve seat with very little effort, reducing heat build up inside
  • Rolled sealing flange provides comfort and improves fitting capabilities
  • Textured sealing area reduces risk of the mask slipping on the face
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Filter Type N95
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small/Medium
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Lacquer, enamel paints, pesticides, welding, dusts
For Use With (Equipment) Cartridges/Diskits
Model 4-SM-20DN5ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.