Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 100 Half-Mask Respirators with OV/R95 Filter Assembly
SDP

Catalog No. 19222435 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Packaging Type:
Display Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-222-435 Small Display Bag
19-222-436 Medium Display Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-222-435 Supplier Dentec Safety™ Supplier No. 1S20DR5ZAM
Add to Cart
Add to Cart

Silicone organic vapor/R95 half-mask respirators are suitable for lacquer and enamel paints, pesticides, welding, dusts, and oil based particulate aerosols for up to eight hours.

  • Crafted from pure silicon material and complete with OV/R95 filter assembly
  • Comfortable and easy to clean
  • Choose from industrial or display bag packaging
  • Low profile design fits under most grinding and welding helmets
  • Extended field of vision
  • Large inhalation valves allow for maximum inhalation
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
  • Offset connector improves visibility and allows face shields or welding helmets to be worn without interference
  • 3 Position adjustment allows users to adjust cartridge to three different positions

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Blue
Filter Type R95
Material Silicone
Product Type Half-Mask Respirator
Size Small
With Exhalation Valve Yes
Facepiece Material Silicone
For Use With (Application) Lacquer, enamel paints, pesticides, welding, dusts
For Use With (Equipment) Cartridges/Diskits
Model 1-S-20DR5ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.