Learn More
Dentec Safety™ Comfort-Ease AD4N95 Flat-Fold N95 Respirators
Shop All Dentec Safety Specialists Inc. Products

Description
- Flat fold design for ideal comfort and fit and allows the user to store the product in a pocket
- Extended field of vision
- Soft material and soft foam seal at nose bridge improves user comfort
- Pass-through straps:
- Allow for quick and simple adjustment
- Allows user to remove crown strap and conveniently hang around the neck
- Ideal for users required to move from a hazardous area to non hazardous area
- Keeps the mask with the user
- Reduces consumption costs
- Adjustable aluminum bar at nose bridge allows user to fit the mask quickly and easily.
- Aluminum bar is sewn into material for dielectric applications
Specifications
Specifications
| Aerosol Type | N - Not Resistant to Oil |
| Certifications (NIOSH) | NIOSH 42 CFR 84, N95 |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | N95 |
| Product Type | General Purpose Particulate Respirator |
| Size | Standard |
| With Exhalation Valve | Yes |
| Filter Efficiency | 95% |
| Latex Free | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.