Learn More
HDS Reusable Comfort Face Masks
Catalog No. 12888102 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-888-102 White
12-888-101 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-888-102
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Mask constructed of antimicrobial fabric with lab results of 99.9% efficacy against bacteria hazardous to upper respiratory health. Speed Tech fabric has fluid repellancy and moisture wicking, allowing for an effective and comfortable mask.
  • Antimicrobial (Microban Z06)
  • Zinc additive
  • Machine washable, tumble dry
  • Efficacy after 50 washes is 99% against 4352 KLEB and 6538 STAPH
  • Water repellency
  • Protects against coughs, sneezes, and other bodily fluids
  • Moisture wicking
  • Combats the condensation build up from breathing
  • Non-stretch fabric
Specifications
Product Type General Purpose Face Mask
Color White
Fastening Type Earloops
Material Polyester
Size Standard
Sterility Non-sterile
Disposable Reusable
Latex Free Yes
Model HDS/2LAYERZINCMASK1/WHITE