Learn More
Med Vet International Comfort Winged Elevator 2.0mm Green, J1231A, Each
Catalog No. 5000583944
Encompass_Preferred
They have short winged blades with lateral edges which curve upwards further than traditional elevators, improving conformation to the root structure as well as instrument control. The narrow neck enhances visibility of the root and the shorter shart improves handling and control, especially if the operator’s hands are not large. The design invorporates the best parts of an elevator and a periodontal knife. Handles are very comfortable in use. Flattened surface prevents rolling off the tables. Available as a boxed set.Comfort Winged Elevator. 2.0mm. Green
Catalog No. 50-005-83944 Supplier Med Vet International Supplier No. JOR1231A
They have short winged blades with lateral edges which curve upwards further than traditional elevators, improving conformation to the root structure as well as instrument control. The narrow neck enhances visibility of the root and the shorter shart improves handling and control, especially if the operator’s hands are not large. The design invorporates the best parts of an elevator and a periodontal knife. Handles are very comfortable in use. Flattened surface prevents rolling off the tables. Available as a boxed set.Comfort Winged Elevator. 2.0mm. Green