Learn More
MSC TPI Commercial Belt Drive Portable Blower, 42 in., 2-Speed, 1/2 hp, 1 ea.
Catalog No. 5000507297
Encompass
This portable commercial belt-drive blower provides high-capacity air circulation for warehouse and industrial environments. Featuring a 2-speed motor, this unit is designed to meet OSHA standards for safety and reliable performance.
  • Features a 42 in. aluminum blade for efficient air movement
  • Constructed with a durable 20-gauge powder-coated steel housing
  • Equipped with a permanently lubricated ball-bearing motor for low maintenance
  • Includes 5 in. rubber wheels for easy portability
  • Features spiral wire front and rear guards for protection
  • Provides 2-speed operation for flexible airflow control
  • Includes an 8 ft. SJT type 3-conductor power cord
  • Meets federal OSHA requirements
Catalog No. 50-005-07297 Supplier MSC Supplier No. 77990554
This portable commercial belt-drive blower provides high-capacity air circulation for warehouse and industrial environments. Featuring a 2-speed motor, this unit is designed to meet OSHA standards for safety and reliable performance.
  • Features a 42 in. aluminum blade for efficient air movement
  • Constructed with a durable 20-gauge powder-coated steel housing
  • Equipped with a permanently lubricated ball-bearing motor for low maintenance
  • Includes 5 in. rubber wheels for easy portability
  • Features spiral wire front and rear guards for protection
  • Provides 2-speed operation for flexible airflow control
  • Includes an 8 ft. SJT type 3-conductor power cord
  • Meets federal OSHA requirements