Learn More
MSC TPI Belt Drive Portable Blower, 36 in., 1/3 HP, 2-Speed, 120V, 1 ea.
Catalog No. 5000507319
Encompass
This 36 in. commercial belt drive portable blower is designed for use in clean, dry environments such as warehouses, shops, offices, and schools. It features a robust 1/3 HP, 2-speed motor and an aluminum blade housed within a durable 20-gauge powder-coated steel frame for efficient air circulation.
  • Features a 2-speed, 1/3 HP, 1-phase motor
  • Equipped with an aluminum blade and spiral wire front and rear guards
  • Built with a 20-gauge powder-coated steel housing
  • Includes 8 in. rubber wheels for portability
  • Features a 120V motor with a rocker switch
  • Meets federal OSHA requirements
  • UL listed for USA and Canada
  • Includes a 1-year limited warranty
Catalog No. 50-005-07319 Supplier MSC Supplier No. 77990547
This 36 in. commercial belt drive portable blower is designed for use in clean, dry environments such as warehouses, shops, offices, and schools. It features a robust 1/3 HP, 2-speed motor and an aluminum blade housed within a durable 20-gauge powder-coated steel frame for efficient air circulation.
  • Features a 2-speed, 1/3 HP, 1-phase motor
  • Equipped with an aluminum blade and spiral wire front and rear guards
  • Built with a 20-gauge powder-coated steel housing
  • Includes 8 in. rubber wheels for portability
  • Features a 120V motor with a rocker switch
  • Meets federal OSHA requirements
  • UL listed for USA and Canada
  • Includes a 1-year limited warranty