Learn More
MSC COMMAND IMPACT C/R GLV
Catalog No. 5000786987
Encompass
These gloves are designed for high-performance protection against cuts, punctures, and abrasion in demanding work environments. They feature an HPPE blend and a foam nitrile-coated palm with a foam grip for enhanced dexterity and impact protection.
  • High-performance protection against cuts (A6), punctures (3), and abrasion (3)
  • Foam nitrile-coated palm with nitrile dots for enhanced grip
  • Touchscreen compatible palm allows use with electronic devices
  • A 13 gauge HPPE blended knit ensures comfort and dexterity
  • Full back-of-hand low-profile TPR impact protection
  • Seamless HPPE lining and knit cuff
Catalog No. 50-007-86987 Supplier MSC Supplier No. 97172167
These gloves are designed for high-performance protection against cuts, punctures, and abrasion in demanding work environments. They feature an HPPE blend and a foam nitrile-coated palm with a foam grip for enhanced dexterity and impact protection.
  • High-performance protection against cuts (A6), punctures (3), and abrasion (3)
  • Foam nitrile-coated palm with nitrile dots for enhanced grip
  • Touchscreen compatible palm allows use with electronic devices
  • A 13 gauge HPPE blended knit ensures comfort and dexterity
  • Full back-of-hand low-profile TPR impact protection
  • Seamless HPPE lining and knit cuff