Learn More
Aviva Systems Biology Corp COMMD1 Antibody - C-terminal region (ARP61706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COMMD1. It was validated on Western Blot. . Alternate names: C2orf5, MGC27155, MURR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-443