Learn More
Aviva Systems Biology Corp COMMD8 Antibody - middle region (ARP57050_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57050_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COMMD8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COMMD8. Alternate names: FLJ20502ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-258