Learn More
Medchemexpress LLC COMMD8 Human N-His 100ug | 100ug
SDP
Catalog No. 5000194008
Encompass_Preferred
COMMD8 Protein Human (N-His) expresses in E coli with a His tag at the N-terminus COMMD8 is a protein that binds to the C-terminal amino acid sequence (residues 306 352) of human CXCR4[1]
Catalog No. 50-001-94008 Supplier Medchemexpress LLC Supplier No. HYP78555100UG
COMMD8 Protein Human (N-His) expresses in E coli with a His tag at the N-terminus COMMD8 is a protein that binds to the C-terminal amino acid sequence (residues 306 352) of human CXCR4[1]