Learn More
MSC Atrix Barrel Vacuum, 55 gal, 3 Motor, HEPA, 1 ea
Catalog No. 502868263
Encompass
The Atrix Barrel Vacuum is designed for applications requiring the collection of large amounts of non-hazardous dry particulate, such as in manufacturing or construction. It features a 3-motor vacuum head that converts a 55-gallon barrel/drum into a high-capacity vacuum, with users able to control suction by activating one, two, or all three motors.
  • Converts 55-gallon barrel/drum into a large capacity vacuum
  • Triple 500 watt motors allow suction control
  • Two-stage HEPA filter configuration
  • HEPA filtration: 99.97% efficient at .3 microns or larger
  • ESD safe
  • Immediate particulate containment
  • Cleanroom compatible
Catalog No. 50-286-8263 Supplier MSC Supplier No. 17758392
The Atrix Barrel Vacuum is designed for applications requiring the collection of large amounts of non-hazardous dry particulate, such as in manufacturing or construction. It features a 3-motor vacuum head that converts a 55-gallon barrel/drum into a high-capacity vacuum, with users able to control suction by activating one, two, or all three motors.
  • Converts 55-gallon barrel/drum into a large capacity vacuum
  • Triple 500 watt motors allow suction control
  • Two-stage HEPA filter configuration
  • HEPA filtration: 99.97% efficient at .3 microns or larger
  • ESD safe
  • Immediate particulate containment
  • Cleanroom compatible