Learn More
Felix Storch Inc Commercial Bullet Ice Machine 55 lb. Bullet Ice Machine production
SDP
Catalog No. NC3901207
Encompass_Preferred
ENERGY STAR certified commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. Inside, the BIM55BU produces up to 55 lbs. of clear, bullet-style ice daily. Bullet ice features a hole at its center, which exposes liquids to more surface area than other ice shapes. The result is a cooler drink in less time than traditional cubed ice. With its unique shape, bullet ice can be produced and harvested fasterAll digital controls are conveniently located at the unit's side, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop.
Catalog No. NC3901207 Supplier Felix Storch Inc Supplier No. BIM55BU
ENERGY STAR certified commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. Inside, the BIM55BU produces up to 55 lbs. of clear, bullet-style ice daily. Bullet ice features a hole at its center, which exposes liquids to more surface area than other ice shapes. The result is a cooler drink in less time than traditional cubed ice. With its unique shape, bullet ice can be produced and harvested fasterAll digital controls are conveniently located at the unit's side, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop.