Learn More
Felix Storch Inc Commercial Bullet Ice Machine 77 lb. clear bullet ice production
SDP
Catalog No. NC3901210
Encompass_Preferred
ENERGY STAR certified, 15 inch wide commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. The BIM77BU produces up to 77 lbs. of clear, bullet-style ice cubes daily. Bullet ice features a hole at its center, which exposes liquids to more surface area than other ice shapes. The result is a cooler drink in less time than traditional cubed ice. The interior can store up to 33 lbs. to help meet the needs of high-traffic areas. All digital controls are conveniently located at the unit's side, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This unit features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop
Catalog No. NC3901210 Supplier Felix Storch Inc Supplier No. BIM77BU
ENERGY STAR certified, 15 inch wide commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. The BIM77BU produces up to 77 lbs. of clear, bullet-style ice cubes daily. Bullet ice features a hole at its center, which exposes liquids to more surface area than other ice shapes. The result is a cooler drink in less time than traditional cubed ice. The interior can store up to 33 lbs. to help meet the needs of high-traffic areas. All digital controls are conveniently located at the unit's side, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This unit features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop