Learn More
Felix Storch Inc Commercial Ice Machine ENERGY STAR certified 120 lb. 80 lb. clear ice production
SDP
Catalog No. NC3901222
Encompass_Preferred
Model BIM85 is an ENERGY STAR certified 18"" deep commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. The BIM85 produces up to 80 lbs. of clear, cube-style ice cubes daily. The interior can store up to 33 lbs. to help meet the needs of high-traffic commercial environments. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop to maximize convenience. The BIM85 is a great fit for break rooms, bars, restaurants, cafes, hotels, and similar spaces.
Catalog No. NC3901222 Supplier Felix Storch Inc Supplier No. BIM85
Model BIM85 is an ENERGY STAR certified 18"" deep commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It is designed for freestanding use and features an air-cooled system to prevent overheating. The BIM85 produces up to 80 lbs. of clear, cube-style ice cubes daily. The interior can store up to 33 lbs. to help meet the needs of high-traffic commercial environments. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop to maximize convenience. The BIM85 is a great fit for break rooms, bars, restaurants, cafes, hotels, and similar spaces.