Learn More
Felix Storch Inc ACCUCOLD Commercial Ice Machine ENERGY STAR certified 120 lb. clear cube-shaped ice production
SDP
Catalog No. NC3901211
Encompass_Preferred
Model BIM125 is an ENERGY STAR certified commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It can be used freestanding with an air-cooled system to prevent overheating. Inside, the BIM125 produces up to 120 lbs. of clear, cube-style ice cubes daily. The interior can store up to 40 lbs. to help meet the needs of high-traffic commercial environments. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop to maximize convenience: https://www.accucold.com/product/BIM125
Catalog No. NC3901211 Supplier Felix Storch Inc Supplier No. BIM125
Model BIM125 is an ENERGY STAR certified commercial ice machine that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It can be used freestanding with an air-cooled system to prevent overheating. Inside, the BIM125 produces up to 120 lbs. of clear, cube-style ice cubes daily. The interior can store up to 40 lbs. to help meet the needs of high-traffic commercial environments. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit also has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop to maximize convenience: https://www.accucold.com/product/BIM125