Learn More
Felix Storch Inc Commercial Ice Machine With Ice Dispenser 350 lb. clear ice production
SDP
Catalog No. NC3901203
Encompass_Preferred
ENERGY STAR certified commercial ice machine with dispenser function that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It can be used freestanding with an air-cooled system to prevent overheating. Inside, the BIM350KDISP produces up to 350 lbs. of clear, cube-style ice cubes daily. The interior can store up to 130 lbs. to help meet the needs of high-traffic commercial environments. It features an ice dispenser which allows for easier access to ice and reduces the risk of contamination by restricting access to the ice storage bin. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop.
Catalog No. NC3901203 Supplier Felix Storch Inc Supplier No. BIM350KDISP
Add to Cart
Add to Cart
Request a Quote
ENERGY STAR certified commercial ice machine with dispenser function that is ETL-S listed to NSF-12 standards. This ice machine features a fully finished stainless steel cabinet and a jet black front with an easy-to-use slide-up lid. It can be used freestanding with an air-cooled system to prevent overheating. Inside, the BIM350KDISP produces up to 350 lbs. of clear, cube-style ice cubes daily. The interior can store up to 130 lbs. to help meet the needs of high-traffic commercial environments. It features an ice dispenser which allows for easier access to ice and reduces the risk of contamination by restricting access to the ice storage bin. All digital controls are conveniently located in front, including a function to adjust the thickness of each ice cube. This unit has a water filter and a removable mesh filter at the rear to ensure easier condenser maintenance. This automatic ice machine features a clean function, a full bin sensor, attractive LED lighting, and an ice scoop.