Learn More
Carolina™ Commercial Sponge Set
SDPSCI_ED
Catalog No. S37718 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
Ea.
Catalog No. Quantity
S37718 Ea.
1 options
Catalog No. S37718 Supplier Carolina™ Supplier No. 224070
Add to Cart
Add to Cart
Carolina™ Commercial Sponge Set is used for examining 4 different dried sponges using this commercial sponge set.
  • Contains 4 dried sponges
  • Each specimen is about 4 in. in diameter
Specifications
Type Preserved Specimen
Preservative Dried
Injection Type Plain
Includes Four sponges: sheep’s wool sponge, yellow sponge, grass sponge, and hard head sponge.
Quantity Ea.
Autoclavable Not Autoclavable