Learn More
Ampco Common Knife
SDP
Catalog No. 19174132 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English) Blade:
3 -1/8 in.
5 -3/4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade
19-174-132 3 -1/8 in.
19-174-136 5 -3/4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-174-132 Supplier Ampco Supplier No. K1
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-reistance knife.
  • Independently tested and found to pose no risk in MRI environments using a static magnetic field of 3-Tesla and a maximum spatial gradient magnetic field of 720-Gauss/cm or less.
  • Conforms to ASTM F2503.
  • Meets the requirements established by ASTM A342.
Specifications
Product Type Common Knife
Length (English) Blade 3 -1/8 in.
Length (English) 6 -3/4 in.
Quantity 1 Ea.