Learn More
MSC Goodway Portable Dry Steam Cleaner, 1650W, 340°F, 105 psi
Catalog No. 502837963
Encompass
This commercial dry steam cleaner is designed for tackling tough cleaning challenges in various environments such as hotels, restaurants, manufacturing plants, and healthcare facilities. It operates without the need for chemicals.
  • 1,650-watt boiler.
  • 340-degree Fahrenheit boiler steam temperature.
  • Up to 105 psi steam pressure.
  • 290°F output steam temperature.
  • 12.5 amps, 115 V, 50/60 Hz AC input power with maximum 1,800 W.
  • 83 inch (211 cm) hose length.
  • 1.3 gallon (5 liter) water tank capacity.
  • 0.6 gallon boiler capacity.
  • Dimensions: 12 inches high x 12 inches wide x 16 inches deep.
  • 15.3 lbs net dry weight.
  • Easy-to-use, plug-and-go operation.
  • Heavy-duty construction.
  • Chemical-free cleaning.
  • Eliminates oil, grease, dirt, and bacteria.
Catalog No. 50-283-7963 Supplier MSC Supplier No. 74247099
This commercial dry steam cleaner is designed for tackling tough cleaning challenges in various environments such as hotels, restaurants, manufacturing plants, and healthcare facilities. It operates without the need for chemicals.
  • 1,650-watt boiler.
  • 340-degree Fahrenheit boiler steam temperature.
  • Up to 105 psi steam pressure.
  • 290°F output steam temperature.
  • 12.5 amps, 115 V, 50/60 Hz AC input power with maximum 1,800 W.
  • 83 inch (211 cm) hose length.
  • 1.3 gallon (5 liter) water tank capacity.
  • 0.6 gallon boiler capacity.
  • Dimensions: 12 inches high x 12 inches wide x 16 inches deep.
  • 15.3 lbs net dry weight.
  • Easy-to-use, plug-and-go operation.
  • Heavy-duty construction.
  • Chemical-free cleaning.
  • Eliminates oil, grease, dirt, and bacteria.