Learn More
Grainger Communication Cable Black 8.2 ft.
Catalog No. 502766131
Encompass_Preferred
The Grainger 16X215 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, rechargeable extinguisher contains 12 lbs of Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment and assets. It features a steel cylinder with red powder coat finish, 12" hose, and pistol grip valve. The extinguisher has a discharge range of 10-15 ft and is for commercial use. It meets NFPA 2010 requirements. The 16X215 model offers an economical, environmentally friendly clean agent fire protection solution.
Catalog No. 50-276-6131 Supplier Grainger Supplier No. 16X215
The Grainger 16X215 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, rechargeable extinguisher contains 12 lbs of Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment and assets. It features a steel cylinder with red powder coat finish, 12" hose, and pistol grip valve. The extinguisher has a discharge range of 10-15 ft and is for commercial use. It meets NFPA 2010 requirements. The 16X215 model offers an economical, environmentally friendly clean agent fire protection solution.