Learn More
Grainger Communication Cable White 39 in
Catalog No. 502765744
Encompass_Preferred
The Schneider Electric 16X213 Communication Cable is a 4-conductor, 24 AWG, stranded copper communication cable with a PVC jacket. It has 4 individually shielded pairs and an overall braided shield. The conductors are color coded. It can be used for RS-232, RS-485, RS-422, Ethernet 10Base-T, and EIA-485 applications. This cable is UL listed and CSA certified. It comes in a box with a 1000 foot spool. The operating temperature range is -20°C to 80°C.
Catalog No. 50-276-5744 Supplier Grainger Supplier No. 16X213
The Schneider Electric 16X213 Communication Cable is a 4-conductor, 24 AWG, stranded copper communication cable with a PVC jacket. It has 4 individually shielded pairs and an overall braided shield. The conductors are color coded. It can be used for RS-232, RS-485, RS-422, Ethernet 10Base-T, and EIA-485 applications. This cable is UL listed and CSA certified. It comes in a box with a 1000 foot spool. The operating temperature range is -20°C to 80°C.