Learn More
Grainger Communication Cable White 98 in
Catalog No. 502766965
Encompass_Preferred
The Schneider Electric Connecting Cable for Altivar M221 Variable Speed Drives is a 3 foot long cable that connects the drive to the motor. It is rated for 600V and designed for variable frequency drive output. The cable has a 16 AWG 3 conductor SEWF flexible cable jacket and is UL and CSA certified. The connecting cable allows quick installation by plugging into the drive and motor.
Catalog No. 50-276-6965 Supplier Grainger Supplier No. 16X216
The Schneider Electric Connecting Cable for Altivar M221 Variable Speed Drives is a 3 foot long cable that connects the drive to the motor. It is rated for 600V and designed for variable frequency drive output. The cable has a 16 AWG 3 conductor SEWF flexible cable jacket and is UL and CSA certified. The connecting cable allows quick installation by plugging into the drive and motor.