Learn More
Sino Biological COMP Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10173T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human COMP (rh COMP; Catalog#10173-H08H; NP_000086.2; Met1-Ala757). COMP specific IgG was purified by Human COMP affinity chromatography.
Catalog No. 50-224-7646