Learn More
Innovative Research Inc Comp C4 Dfcnt Hmn Pla Lyo 1ml
Catalog No. 502744616
Encompass_Preferred
Complement C4 Deficient Human Plasma Lyophilized 1ml - Complement C4 Deficient Human Plasma from Innovative Research is supplied as a vial of depleted human plasma in lyophilized form. The plasma was established as 99% deficient on average by ELISA testing. Albumin level serves as a control for the depletion process.
Catalog No. 50-274-4616 Supplier Innovative Research Inc Supplier No. IC4DPLY1ML
Complement C4 Deficient Human Plasma Lyophilized 1ml - Complement C4 Deficient Human Plasma from Innovative Research is supplied as a vial of depleted human plasma in lyophilized form. The plasma was established as 99% deficient on average by ELISA testing. Albumin level serves as a control for the depletion process.