Learn More
AdipoGen COMP ratAng1 h rec
SDP
Catalog No. 502562911
Encompass_Preferred
Protein. The fibrinogen-like domain aa 284-498 of human angiopoietin-1 is fused at the N-terminus to the coiled-coil domain of rat COMP cartilage oligomeric matrix protein and to a FLAG -tag. Source CHO cells. Endotoxin content <0.05EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Binds to human angiopoietin-1 receptor Tie-2. Purity >95% SDS-PAGE. Soluble, stable and more potent angiopoietin-1 Ang-1 variant.
Catalog No. 50-256-2911 Supplier AdipoGen Supplier No. AG40B0147C010
Protein. The fibrinogen-like domain aa 284-498 of human angiopoietin-1 is fused at the N-terminus to the coiled-coil domain of rat COMP cartilage oligomeric matrix protein and to a FLAG -tag. Source CHO cells. Endotoxin content <0.05EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Binds to human angiopoietin-1 receptor Tie-2. Purity >95% SDS-PAGE. Soluble, stable and more potent angiopoietin-1 Ang-1 variant.