Learn More
AdipoGen COMP (rat):Angiopoietin-1 (human) (rec.) (5 x 10 g)
SDP
Supplier:  AdipoGen AG40B01475010
Encompass_Preferred
COMP (rat):Angiopoietin-1 (human) (rec.) (5 x 10 g), AdipoGen Life Sciences. Protein. The fibrinogen-like domain (aa 284-498) of human angiopoietin-1 is fused at the N-terminus to the coiled-coil domain of rat COMP (cartilage oligomeric matrix protein) and to a FLAG-tag. Source: CHO cells. Endotoxin content: <0.05EU/g purified protein (LAL test; Lonza). Lyophilized. Contains PBS + 0.02% CHAPS. Binds to human angiopoietin-1 receptor (Tie-2). Purity: >95% (SDS-PAGE). Soluble, stable and more potent angiopoietin-1 (Ang-1) variant.
Catalog No. 50-148-9431