Learn More
Waters Corp WATERS CORPORATION Compression Screw, 1/16 in, 10/pk
Catalog No. 50819009 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters™ tubing and fittings are designed to ensure leak-free connections, low dead volume, and minimal band spreading. These components are engineered for optimal performance, requiring precise matching between the ferrule geometry and conical port, as well as the tube and fitting diameters. A diverse range of materials, lengths, and diameters offers flexibility for system setup and ongoing maintenance.
  • Designed to provide leak-free connections
  • Features low dead volume
  • Ensures minimal band spreading
  • Engineered for optimal performance
  • Requires matching ferrule and conical port geometry
  • Requires matching tube and fitting diameters
  • Available in a variety of materials, lengths, and diameters
  • Offers flexibility for system setup and maintenance
Catalog No. 50-819-009 Supplier Waters Corp Supplier No. WAT005070
Waters™ tubing and fittings are designed to ensure leak-free connections, low dead volume, and minimal band spreading. These components are engineered for optimal performance, requiring precise matching between the ferrule geometry and conical port, as well as the tube and fitting diameters. A diverse range of materials, lengths, and diameters offers flexibility for system setup and ongoing maintenance.
  • Designed to provide leak-free connections
  • Features low dead volume
  • Ensures minimal band spreading
  • Engineered for optimal performance
  • Requires matching ferrule and conical port geometry
  • Requires matching tube and fitting diameters
  • Available in a variety of materials, lengths, and diameters
  • Offers flexibility for system setup and maintenance