Learn More
Alpco Ltd Complement System Alternative Pathway ELISA
SDP
Supplier:  Alpco Ltd 13COMPLAP330
Encompass
The Complement System Alternative Pathway ELISA kit is an enzyme immunoassay for the qualitative determination of functional alternative complement pathway in human serum. For Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC1286173