Learn More
Alkali Scientific Alkali Scientific™ Compact Anti Static Ionizer for Analytical Balance
SDP
Catalog No. 503029584
Encompass_Preferred
Alkali Scientific Anti Static Ionizer Static charge can accumulate on many types of samples as well as weighing containers, affecting mass determination on an analytical balance.  The Alkali Scientific ASI-100 produces and emits negatively and positively charged ions that will neutralize static electricity on samples and containers.  A blower can be turned on for rapid static discharge, or turned off for work with powders or light-weight materials.
Catalog No. 50-302-9584 Supplier Alkali Scientific Supplier No. W3120
Alkali Scientific Anti Static Ionizer Static charge can accumulate on many types of samples as well as weighing containers, affecting mass determination on an analytical balance.  The Alkali Scientific ASI-100 produces and emits negatively and positively charged ions that will neutralize static electricity on samples and containers.  A blower can be turned on for rapid static discharge, or turned off for work with powders or light-weight materials.