Learn More
Rice Lake Weighing System Equipoise; Series Compact Balance 3000g x 0.1g
Catalog No. 501542694
Encompass_Preferred
Ideal for classrooms, laboratories and industrial applications. These seven-button balances allow quick access to 13 different units of measure, percentage, counting and accumulating. Equipoise can be used with the included AC adapter or an on-board rechargeable battery that offers up to 40 hours of continuous operation. RS-232 output is standard on each model. RS-232 cable is optional. Equipoise Compact Balance 3000g x 0.1g internal resolution 1/600,000, LCD Display , Accumulation counting and percentage modes, Standard RS-232 interface, 13 different units of measure, AC Adapter w/ internal rechargeable battery
Catalog No. 50-154-2694 Supplier Rice Lake Weighing System Supplier No. 165341
Ideal for classrooms, laboratories and industrial applications. These seven-button balances allow quick access to 13 different units of measure, percentage, counting and accumulating. Equipoise can be used with the included AC adapter or an on-board rechargeable battery that offers up to 40 hours of continuous operation. RS-232 output is standard on each model. RS-232 cable is optional. Equipoise Compact Balance 3000g x 0.1g internal resolution 1/600,000, LCD Display , Accumulation counting and percentage modes, Standard RS-232 interface, 13 different units of measure, AC Adapter w/ internal rechargeable battery