Learn More
Rice Lake Weighing System Equipoise; Series Compact Balance 600g x 0.02g includes weighing chamber
Catalog No. 501542697
Encompass_Preferred
Ideal for classrooms, laboratories and industrial applications. These seven-button balances allow quick access to 13 different units of measure, percentage, counting and accumulating. A detachable enclosed weighing chamber is included in capacities below 1000 grams. Equipoise can be used with the included AC adapter or an on-board rechargeable battery that offers up to 40 hours of continuous operation. RS-232 output is standard on each model. RS-232 cable is optional. Equipoise Compact Balance 600g x 0.02g internal resolution 1/600,000, LCD Display , Accumulation counting and percentage modes, Standard RS-232 interface, 13 different units of measure, AC Adapter w/ internal rechargeable battery
Catalog No. 50-154-2697 Supplier Rice Lake Weighing System Supplier No. 165339
Ideal for classrooms, laboratories and industrial applications. These seven-button balances allow quick access to 13 different units of measure, percentage, counting and accumulating. A detachable enclosed weighing chamber is included in capacities below 1000 grams. Equipoise can be used with the included AC adapter or an on-board rechargeable battery that offers up to 40 hours of continuous operation. RS-232 output is standard on each model. RS-232 cable is optional. Equipoise Compact Balance 600g x 0.02g internal resolution 1/600,000, LCD Display , Accumulation counting and percentage modes, Standard RS-232 interface, 13 different units of measure, AC Adapter w/ internal rechargeable battery