Learn More
STEMCELL Technologies PBS-MINI 0.1 MAG Single-Use Vessel, Size: 4 Units
SDP
Catalog No. 502250337
Encompass
Reliably and rapidly scale up your 3D cell cultures and suspensions with the PBS-MINI MagDrive Bioreactor Base Unit and MAG Single-Use Vessels. The gentle yet efficient mixing provided by the Vertical-Wheel™ impeller enables the expansion of shear-sensitive cells without anti-foaming agents or shear protectants. Ideal for human pluripotent stem cells (hPSCs), the compact, sealed Base Unit can be used inside incubators. Conveniently control your culture system with a speed dial and digital display, and visualize cells in low-light conditions using built-in LED lights. This Base Unit is designed to be used with the 0.1 L and 0.5 L PBS-MINI MAG Single-Use Vessels, which it magnetically drives via MagDrive technology. These animal component-free disposable vessels are sold separately in packs of 4. To learn more about the PBS-MINI Vertical-Wheel™ Bioreactor, visit www.stemcell.com/pbs-mini. This product is for Research Use Only.
This item is not returnable. View return policy
Catalog No. 50-225-0337 Supplier STEMCELL Technologies Supplier No. 1001006
Reliably and rapidly scale up your 3D cell cultures and suspensions with the PBS-MINI MagDrive Bioreactor Base Unit and MAG Single-Use Vessels. The gentle yet efficient mixing provided by the Vertical-Wheel™ impeller enables the expansion of shear-sensitive cells without anti-foaming agents or shear protectants. Ideal for human pluripotent stem cells (hPSCs), the compact, sealed Base Unit can be used inside incubators. Conveniently control your culture system with a speed dial and digital display, and visualize cells in low-light conditions using built-in LED lights. This Base Unit is designed to be used with the 0.1 L and 0.5 L PBS-MINI MAG Single-Use Vessels, which it magnetically drives via MagDrive technology. These animal component-free disposable vessels are sold separately in packs of 4. To learn more about the PBS-MINI Vertical-Wheel™ Bioreactor, visit www.stemcell.com/pbs-mini. This product is for Research Use Only.