Learn More
Med Vet International Compact Blue Foam 3/8” Thick Padding (for Splints), 16" x 24" Sheet, Blue, J0940X, Each
Catalog No. 5000581168
Encompass_Preferred
With self-adhesive back. Open-cell, slow-recovery construction. Medium-density, compact foam molds with body heat to contour body parts. Use to pad the splint for increased patient comfort. Contains some latex.
Catalog No. 50-005-81168 Supplier Med Vet International Supplier No. JOR940X
With self-adhesive back. Open-cell, slow-recovery construction. Medium-density, compact foam molds with body heat to contour body parts. Use to pad the splint for increased patient comfort. Contains some latex.