Learn More
Adam Equipment CQT 251 Core Compact Portable Precision Balance, 250g Capacity and 0.1g Readability
Catalog No. NC0561567
Encompass_Preferred
Adam Equipments CQT Core compact balances offers outstanding value, providing easy operation and a portable solution for any basic precision weighing tasks. An innovative stackable design saves space and can easily be stored in school supply cupboards and prep rooms until needed, ideal for busy science classrooms or laboratory environments. Offering precision results, Core is suitable for weighing compounds in science classrooms, samples at a field lab, ingredients in food labs and specimens at veterinary offices. The Core includes a convenient weigh-below hook, facilitating below-balance weighing for tasks requiring density and specific gravity measurements.Hook included for weighing below the balance. Operates with AC adapter (included) or batteries (not included).
Catalog No. NC0561567 Supplier Adam Equipment Supplier No. CQT 251
Adam Equipments CQT Core compact balances offers outstanding value, providing easy operation and a portable solution for any basic precision weighing tasks. An innovative stackable design saves space and can easily be stored in school supply cupboards and prep rooms until needed, ideal for busy science classrooms or laboratory environments. Offering precision results, Core is suitable for weighing compounds in science classrooms, samples at a field lab, ingredients in food labs and specimens at veterinary offices. The Core includes a convenient weigh-below hook, facilitating below-balance weighing for tasks requiring density and specific gravity measurements.Hook included for weighing below the balance. Operates with AC adapter (included) or batteries (not included).