Learn More
Cole-Parmer Soleus HCT-D45E-A Compact Digital Dehumidifier, 45 Pints; 120 V, 60 Hz
Catalog No. NC0710690 Shop All Cole-Parmer Products
Encompass
These dehumidifiers feature three humidity-level settings, three fan-speed settings, continuous dehumidification, full-tank indicator, and a water drain for continuous operation. A magnetic float within the water tank provides accurate water level readings and prevents tank overflow. When full, the tank indicator light illuminates and the unit automatically shuts off. Selectable humidity levels offer options for any comfort level. Once the selected humidity level is achieved, the compressor shuts off automatically. In situations where constant dehumidification is necessary, the continuous operation mode overrides the level setting. For convenient portability, the units feature a carrying handle and casters.
Catalog No. NC0710690 Supplier Cole-Parmer Supplier No. 3770111
These dehumidifiers feature three humidity-level settings, three fan-speed settings, continuous dehumidification, full-tank indicator, and a water drain for continuous operation. A magnetic float within the water tank provides accurate water level readings and prevents tank overflow. When full, the tank indicator light illuminates and the unit automatically shuts off. Selectable humidity levels offer options for any comfort level. Once the selected humidity level is achieved, the compressor shuts off automatically. In situations where constant dehumidification is necessary, the continuous operation mode overrides the level setting. For convenient portability, the units feature a carrying handle and casters.