Learn More
Research Products International Corp Stackable Compact Desiccator Cabinet
SDP
Catalog No. 50212961
Encompass
This cabinet is injection molded of high impact strength polystyrene for attractive styling and rugged durability. The full transparency of this material allows total visibility of contents from all sides. A foam rubber gasket provides an airtight seal when the door is closed and latched. The door stores in a convenient underneath compartment when opened.
The cabinet is designed for safe and convenient stacking up to five units high.
Each cabinet is packaged with a supply of silica gel adsorbent to extract moisture from the air present within the cabinet.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-961 Supplier Research Products International Corp Supplier No. H42053
This cabinet is injection molded of high impact strength polystyrene for attractive styling and rugged durability. The full transparency of this material allows total visibility of contents from all sides. A foam rubber gasket provides an airtight seal when the door is closed and latched. The door stores in a convenient underneath compartment when opened.
The cabinet is designed for safe and convenient stacking up to five units high.
Each cabinet is packaged with a supply of silica gel adsorbent to extract moisture from the air present within the cabinet.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.