Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Compact Digital Dry Bath/Block Heater
SCI_ED

Catalog No. S29052 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Compact Dry Bath D
Compact Dry Bath S
Dimensions (L x W x H):
185 x 185 x 25 mm
200 x 200 x 25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Dimensions (L x W x H)
S2-905-2 Compact Dry Bath D 200 x 200 x 25 mm
S2-905-0 Compact Dry Bath S 185 x 185 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S2-905-2 Supplier Thermo Scientific™ Supplier No. 88871002/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.

Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.

  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification

Required Accessory:

  • Unit must be ordered with block(s)

Warranty and Services

2 Year, Replacement

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cTUVus, RoHS
Controller Type Digital
Plug Type US
Temperature Range (Metric) Ambient +2° to 100°C
Type Dry Block Heater
Heating Rate 20 min. (30° to 100°C, ambient at 25°C, with one 0.2 mL block)
Includes Dry Bath, US style plug, block lifter, plastic lid
Model Compact Dry Bath D
No. of Blocks 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 0.5°C
Warranty 2 Year, Replacement
Dimensions (L x W x H) 200 x 200 x 25 mm
Length (English) 7.9 in.
Length (Metric) 200 mm
Width (English) 7.9 in.
Width (Metric) 200 mm
Height (English) 1 in.
Height (Metric) 25 mm
Weight (English) 3.3 lb.
Weight (Metric) <1.5 kg
Electrical Requirements 12 V, DC, 7 A
Voltage 100/240 V
Show More Show Less

Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.